Learn More
Medchemexpress LLC DLin-KC2-DMA | 1190197-97-7 | ≥98.0% | 642.09 | 25 MG
SDP
Catalog No. 5000439596
Encompass_Preferred
DLin-KC2-DMA is an ionizable cationic lipid (pKa≈6) that is virtually non-toxic to antigen presenting cells (APCs). It produces significant siRNA-mediated gene silencing of GAPDH when bound to lipid nanoparticles (LNP). DLin-KC2-DMA can be used in siRNA delivery studies.
  • Ionizable cationic lipid (pKa≈6).
  • Virtually non-toxic to antigen presenting cells (APCs).
  • Produces significant siRNA-mediated gene silencing of GAPDH when bound to lipid nanoparticles (LNP).
  • Can be used in siRNA delivery studies.
  • High uptake into macrophages and dendritic cells.
  • Efficiently promotes release of encapsulated siRNA into the cytosol following uptake via the endocytotic pathway.
  • Displays almost no toxicity in primary APCs.
  • Effective in silencing target genes in APCs in vivo (5 mg/kg; i.v.; single).
Catalog No. 50-004-39596 Supplier Medchemexpress LLC Supplier No. 25MGHY112758
DLin-KC2-DMA is an ionizable cationic lipid (pKa≈6) that is virtually non-toxic to antigen presenting cells (APCs). It produces significant siRNA-mediated gene silencing of GAPDH when bound to lipid nanoparticles (LNP). DLin-KC2-DMA can be used in siRNA delivery studies.
  • Ionizable cationic lipid (pKa≈6).
  • Virtually non-toxic to antigen presenting cells (APCs).
  • Produces significant siRNA-mediated gene silencing of GAPDH when bound to lipid nanoparticles (LNP).
  • Can be used in siRNA delivery studies.
  • High uptake into macrophages and dendritic cells.
  • Efficiently promotes release of encapsulated siRNA into the cytosol following uptake via the endocytotic pathway.
  • Displays almost no toxicity in primary APCs.
  • Effective in silencing target genes in APCs in vivo (5 mg/kg; i.v.; single).