Learn More
Cayman Chemical DLin-KC2-DMA, Purity: ≥98%, CAS: 1190197-97-7, 5 mg
SDP
Catalog No. 502949051
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.7); has been used in the formation of LNPs encapsulating siRNA or plasmid DNA for use in vitro and in vivo; LNPs containing DLin-KC2-DMA selectively accumulate in the liver after i.v. or i.m. administration in mice or localize to the retinal pigment epithelium and Müller glia after subretinal administration; DLin-KC2-DMA-containing LNPs encapsulating F7-targeted siRNA reduce serum Factor VII levels in mice; DLin-KC2-DMA-containing LNPs encapsulating AR-targeted siRNA decrease serum PSA and tumor androgen receptor levels in an LNCaP prostate cancer mouse xenograft model
Catalog No. 50-294-9051 Supplier Cayman Chemical Supplier No. 343635 MG
An ionizable cationic lipid (pKa = 6.7); has been used in the formation of LNPs encapsulating siRNA or plasmid DNA for use in vitro and in vivo; LNPs containing DLin-KC2-DMA selectively accumulate in the liver after i.v. or i.m. administration in mice or localize to the retinal pigment epithelium and Müller glia after subretinal administration; DLin-KC2-DMA-containing LNPs encapsulating F7-targeted siRNA reduce serum Factor VII levels in mice; DLin-KC2-DMA-containing LNPs encapsulating AR-targeted siRNA decrease serum PSA and tumor androgen receptor levels in an LNCaP prostate cancer mouse xenograft model