Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DLin-M-C4-DMA | 1226909-66-5 | C44H81NO2 | 100 MG
SDP

Catalog No. 5000376925
Encompass_Preferred

DLin-M-C4-DMA is a cationic lipid (Compound MC4) that can be utilized for the delivery of nucleic acids.

  • Cationic lipid
  • Utilized for nucleic acid delivery
  • Purity 98.0%
  • Molecular weight 656.12
  • Chemical formula C44H81NO2
  • Appears as a liquid
  • Colorless to light yellow appearance

Catalog No. 50-003-76925 Supplier Medchemexpress LLC Supplier No. HY154804100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-C4-DMA is a cationic lipid (Compound MC4) that can be utilized for the delivery of nucleic acids.

  • Cationic lipid
  • Utilized for nucleic acid delivery
  • Purity 98.0%
  • Molecular weight 656.12
  • Chemical formula C44H81NO2
  • Appears as a liquid
  • Colorless to light yellow appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.