Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DLin-M-C4-DMA | 1226909-66-5 | C44H81NO2 | 5 MG
SDP

Catalog No. 5000376926
Encompass_Preferred

DLin-M-C4-DMA (Compound MC4) is a cationic lipid used for the delivery of nucleic acids.

  • Cationic lipid
  • Used for nucleic acid delivery
  • Molecular weight: 656.12
  • Appearance: Colorless to light yellow liquid
  • Density: 0.886 g/cm³

Catalog No. 50-003-76926 Supplier Medchemexpress LLC Supplier No. HY1548045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-C4-DMA (Compound MC4) is a cationic lipid used for the delivery of nucleic acids.

  • Cationic lipid
  • Used for nucleic acid delivery
  • Molecular weight: 656.12
  • Appearance: Colorless to light yellow liquid
  • Density: 0.886 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.