Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DLin-M-K-DMA | 1221271-55-1 | 98.0% | 628.07 g·mol⁻1 | C42H77NO2 | 10 MG
SDP

Catalog No. 5000285957
Encompass_Preferred

DLin-M-K-DMA (DLin-M-C2-DMA) is an ionizable cationic lipid supplied as a high-purity liquid for research use in lipid nanoparticle and other nucleic acid delivery formulations. It is commonly used to formulate LNPs for siRNA and mRNA delivery and includes solubility and storage guidance for in vitro and in vivo applications.

  • High purity (98.0%) suitable for formulation and research.
  • Ionizable cationic lipid for nucleic acid delivery applications.
  • Supplied as a liquid, colorless to light yellow.
  • Soluble in DMSO at 100 mg/mL; compatible with common in vivo vehicles.
  • Storage recommendations provided for long-term and solvent conditions.
  • Suitable for incorporation into lipid nanoparticle formulation workflows.

Catalog No. 50-002-85957 Supplier Medchemexpress LLC Supplier No. HY15480310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-K-DMA (DLin-M-C2-DMA) is an ionizable cationic lipid supplied as a high-purity liquid for research use in lipid nanoparticle and other nucleic acid delivery formulations. It is commonly used to formulate LNPs for siRNA and mRNA delivery and includes solubility and storage guidance for in vitro and in vivo applications.

  • High purity (98.0%) suitable for formulation and research.
  • Ionizable cationic lipid for nucleic acid delivery applications.
  • Supplied as a liquid, colorless to light yellow.
  • Soluble in DMSO at 100 mg/mL; compatible with common in vivo vehicles.
  • Storage recommendations provided for long-term and solvent conditions.
  • Suitable for incorporation into lipid nanoparticle formulation workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.