Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dlin-m-k-dma | 1221271-55-1 | C42H77NO2 | 1 ML
SDP

Catalog No. 5000376049
Encompass_Preferred

DLin-M-K-DMA is a cationic lipid used in the construction of biological delivery vectors. It is intended for research use only.

  • Purity of 98.0%.
  • Molecular weight of 628.07.
  • Chemical formula C42H77NO2.
  • CAS number 1221271-55-1.
  • Appears as a liquid, colorless to light yellow.
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-003-76049 Supplier Medchemexpress LLC Supplier No. HY15480310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-K-DMA is a cationic lipid used in the construction of biological delivery vectors. It is intended for research use only.

  • Purity of 98.0%.
  • Molecular weight of 628.07.
  • Chemical formula C42H77NO2.
  • CAS number 1221271-55-1.
  • Appears as a liquid, colorless to light yellow.
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.