Learn More
Medchemexpress LLC DLin-M-K-DMA | 1221271-55-1 | C42H77NO2 | 100 MG
SDP
Catalog No. 5000376462
Encompass_Preferred
DLin-M-K-DMA, also known as DLin-M-C2-DMA, is a cationic lipid primarily utilized in the creation of biological delivery vectors. This compound is intended for research purposes only and is not sold for direct patient use.
  • Cationic lipid.
  • Used in the construction of biological delivery vectors.
  • High purity, specified at 98.0%.
  • Available in various forms including liquid and solutions.
  • Pure form storage: -20°C for 3 years or 4°C for 2 years.
  • In solvent storage: -80°C for 6 months or -20°C for 1 month.
  • Different dissolution protocols are available for in vivo experiments, offering clear or suspended solutions.
Catalog No. 50-003-76462 Supplier Medchemexpress LLC Supplier No. HY154803100MG
DLin-M-K-DMA, also known as DLin-M-C2-DMA, is a cationic lipid primarily utilized in the creation of biological delivery vectors. This compound is intended for research purposes only and is not sold for direct patient use.
  • Cationic lipid.
  • Used in the construction of biological delivery vectors.
  • High purity, specified at 98.0%.
  • Available in various forms including liquid and solutions.
  • Pure form storage: -20°C for 3 years or 4°C for 2 years.
  • In solvent storage: -80°C for 6 months or -20°C for 1 month.
  • Different dissolution protocols are available for in vivo experiments, offering clear or suspended solutions.