Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DLin-M-K-DMA | 1221271-55-1 | C42H77NO2 | 5 MG
SDP

Catalog No. 5000376594
Encompass_Preferred

DLin-M-K-DMA is a cationic lipid that can be used in the construction of biological delivery vectors. This compound is provided as a colorless to light yellow liquid with a purity of 98.0%.

  • Cationic lipid
  • Suitable for biological delivery vectors
  • Molecular weight: 628.07
  • Chemical formula: C42H77NO2
  • CAS number: 1221271-55-1
  • Appearance: Colorless to light yellow liquid
  • Purity: 98.0%

Catalog No. 50-003-76594 Supplier Medchemexpress LLC Supplier No. HY1548035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-K-DMA is a cationic lipid that can be used in the construction of biological delivery vectors. This compound is provided as a colorless to light yellow liquid with a purity of 98.0%.

  • Cationic lipid
  • Suitable for biological delivery vectors
  • Molecular weight: 628.07
  • Chemical formula: C42H77NO2
  • CAS number: 1221271-55-1
  • Appearance: Colorless to light yellow liquid
  • Purity: 98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.