Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DLin-M-K-DMA (DLin-M-C2-DMA) | 1221271-55-1 | C42H77NO2 | 250 MG
SDP

Catalog No. 5000374291
Encompass_Preferred

DLin-M-K-DMA (DLin-M-C2-DMA) is a cationic lipid that can be used in the construction of biological delivery vectors.

  • Cationic lipid for biological delivery applications.
  • Purity of 98.0%.
  • Molecular weight of 628.07.
  • Appears as a colorless to light yellow liquid.
  • Soluble in DMSO at 100 mg/mL.
  • Stable for up to 3 years in pure form at -20°C.
  • Stable for up to 6 months in solvent at -80°C.

Catalog No. 50-003-74291 Supplier Medchemexpress LLC Supplier No. 250MGHY154803
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-K-DMA (DLin-M-C2-DMA) is a cationic lipid that can be used in the construction of biological delivery vectors.

  • Cationic lipid for biological delivery applications.
  • Purity of 98.0%.
  • Molecular weight of 628.07.
  • Appears as a colorless to light yellow liquid.
  • Soluble in DMSO at 100 mg/mL.
  • Stable for up to 3 years in pure form at -20°C.
  • Stable for up to 6 months in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.