Learn More
Medchemexpress LLC DLin-M-K-DMA (DLin-M-C2-DMA) | 1221271-55-1 | C42H77NO2 | 50 MG
SDP
Catalog No. 5000376181
Encompass_Preferred
DLin-M-K-DMA (DLin-M-C2-DMA) is a cationic lipid primarily utilized in the construction of biological delivery vectors. It is suitable for various research applications requiring efficient gene or drug delivery systems. The product should be stored carefully to maintain its stability and efficacy over time.
  • Cationic lipid for biological delivery systems
  • Used in the construction of delivery vectors
  • For research use only
  • Store at -20°C for up to 1 month (stock solution)
  • Store at -80°C for up to 6 months (stock solution)
  • Keep away from direct sunlight and sources of ignition
Catalog No. 50-003-76181 Supplier Medchemexpress LLC Supplier No. HY15480350MG
DLin-M-K-DMA (DLin-M-C2-DMA) is a cationic lipid primarily utilized in the construction of biological delivery vectors. It is suitable for various research applications requiring efficient gene or drug delivery systems. The product should be stored carefully to maintain its stability and efficacy over time.
  • Cationic lipid for biological delivery systems
  • Used in the construction of delivery vectors
  • For research use only
  • Store at -20°C for up to 1 month (stock solution)
  • Store at -80°C for up to 6 months (stock solution)
  • Keep away from direct sunlight and sources of ignition