Learn More
Cayman Chemical DLIn-MC2-DMA 10mg
SDP
Catalog No. 502945500
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs; LNPs containing DLin-MC2-DMA and encapsulating siRNA targeting mRNA encoding FVII reduce hepatic levels of FVII in mice (ED50 = 0.6 mg/kg)
Catalog No. 50-294-5500 Supplier Cayman Chemical Supplier No. 3973110 MG
An ionizable cationic lipid; has been used in the formation of LNPs; LNPs containing DLin-MC2-DMA and encapsulating siRNA targeting mRNA encoding FVII reduce hepatic levels of FVII in mice (ED50 = 0.6 mg/kg)