Learn More
Cayman Chemical DLin-MC2-DMA, Purity: ≥95%, CAS: 1221271-55-1, 25 mg
SDP
Catalog No. 502943290
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs; LNPs containing DLin-MC2-DMA and encapsulating siRNA targeting mRNA encoding FVII reduce hepatic levels of FVII in mice (ED50 = 0.6 mg/kg)
Catalog No. 50-294-3290 Supplier Cayman Chemical Supplier No. 3973125 MG
An ionizable cationic lipid; has been used in the formation of LNPs; LNPs containing DLin-MC2-DMA and encapsulating siRNA targeting mRNA encoding FVII reduce hepatic levels of FVII in mice (ED50 = 0.6 mg/kg)