Learn More
Cayman Chemical DLin-MC2-DMA, Purity: ≥95%, CAS: 1221271-55-1, 5 mg
SDP
Catalog No. 502944788
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs; LNPs containing DLin-MC2-DMA and encapsulating siRNA targeting mRNA encoding FVII reduce hepatic levels of FVII in mice (ED50 = 0.6 mg/kg)
Catalog No. 50-294-4788 Supplier Cayman Chemical Supplier No. 397315 MG
An ionizable cationic lipid; has been used in the formation of LNPs; LNPs containing DLin-MC2-DMA and encapsulating siRNA targeting mRNA encoding FVII reduce hepatic levels of FVII in mice (ED50 = 0.6 mg/kg)