Learn More
Cayman Chemical DLin-MC3-DMA, Purity: ≥95%, CAS: 1224606-06-7, 10 mg
SDP
Catalog No. 502949813
Encompass_Preferred
An ionizable cationic lipid (apparent pKa = 6.44); has been used in the generation of lipid nanoparticles (LNPs) encapsulating siRNA, mRNA, or plasmid DNA for use in vitro and in vivo; LNPs containing DLin-MC3-DMA accumulate in the muscle and non-draining lymph nodes after intramuscular administration and in the spleen after intravenous administration in mice; LNPs containing DLin-MC3-DMA and encapsulating F7 siRNA reduce hepatic and plasma Factor VII levels in mice without inducing liver toxicity
Catalog No. 50-294-9813 Supplier Cayman Chemical Supplier No. 3436410 MG
An ionizable cationic lipid (apparent pKa = 6.44); has been used in the generation of lipid nanoparticles (LNPs) encapsulating siRNA, mRNA, or plasmid DNA for use in vitro and in vivo; LNPs containing DLin-MC3-DMA accumulate in the muscle and non-draining lymph nodes after intramuscular administration and in the spleen after intravenous administration in mice; LNPs containing DLin-MC3-DMA and encapsulating F7 siRNA reduce hepatic and plasma Factor VII levels in mice without inducing liver toxicity