Learn More
Cayman Chemical DLIn-MC4-DMA 100mg
SDP
Catalog No. 502943031
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.93); LNPs containing DLin-MC4-DMA and encapsulating siRNA for F7 reduce its gene expression levels in mice
Catalog No. 50-294-3031 Supplier Cayman Chemical Supplier No. 39732100 MG
An ionizable cationic lipid (pKa = 6.93); LNPs containing DLin-MC4-DMA and encapsulating siRNA for F7 reduce its gene expression levels in mice