Learn More
Cayman Chemical DLin-MC4-DMA, Purity: ≥95%, CAS: 1226909-66-5, 10 mg
SDP
Catalog No. 502945501
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.93); LNPs containing DLin-MC4-DMA and encapsulating siRNA for F7 reduce its gene expression levels in mice
Catalog No. 50-294-5501 Supplier Cayman Chemical Supplier No. 3973210 MG
An ionizable cationic lipid (pKa = 6.93); LNPs containing DLin-MC4-DMA and encapsulating siRNA for F7 reduce its gene expression levels in mice