Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dlin-MeOH | 1169768-28-8 | 98.0% | C37H68O | 50 MG
SDP

Catalog No. 5000354698
Encompass_Preferred

Dlin-MeOH is a lipid product intended for use in drug delivery systems. It is for research use only and not sold to patients. The appearance is a liquid with a colorless to light yellow color, and a density of 0.864 ± 0.06 g/cm3. It is soluble in DMSO at 100 mg/mL (189.06 mM) and requires ultrasonication for dissolution.

  • Purity of 98.0%
  • Molecular weight of 528.94
  • Chemical formula: C37H68O
  • Intended for research use only
  • Used in drug delivery systems

Catalog No. 50-003-54698 Supplier Medchemexpress LLC Supplier No. HY14298650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dlin-MeOH is a lipid product intended for use in drug delivery systems. It is for research use only and not sold to patients. The appearance is a liquid with a colorless to light yellow color, and a density of 0.864 ± 0.06 g/cm3. It is soluble in DMSO at 100 mg/mL (189.06 mM) and requires ultrasonication for dissolution.

  • Purity of 98.0%
  • Molecular weight of 528.94
  • Chemical formula: C37H68O
  • Intended for research use only
  • Used in drug delivery systems

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.