Learn More
Medchemexpress LLC Dlin-MeOH | 1169768-28-8 | MFCD32062843 | 98.0% | C37H68O | 100 MG
SDP
Catalog No. 5000355332
Encompass_Preferred
Dlin-MeOH is a lipid product primarily used in drug delivery systems. It also serves as a versatile laboratory chemical for the synthesis and manufacture of various substances.
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month.
  • Shipping at room temperature if less than 2 weeks.
  • Keep container tightly sealed in cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.
Catalog No. 50-003-55332 Supplier Medchemexpress LLC Supplier No. HY142986100MG
Dlin-MeOH is a lipid product primarily used in drug delivery systems. It also serves as a versatile laboratory chemical for the synthesis and manufacture of various substances.
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month.
  • Shipping at room temperature if less than 2 weeks.
  • Keep container tightly sealed in cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.