Learn More
Medchemexpress LLC DLinDMA | 871258-12-7 | 99.49% | 616.06 | 100 MG
SDP
Catalog No. 5000437220
Encompass_Preferred
DLinDMA, an ionizable cationic lipid, is a key lipid component of stable nucleic acid lipid particles (SNALPs) as a benchmark. It is used for siRNA delivery. Its structure can be divided into three main regions: the hydrocarbon chains, the linker, and the headgroup.
  • Ionizable cationic lipid
  • Key lipid component of stable nucleic acid lipid particles (SNALPs)
  • Used for siRNA delivery
Catalog No. 50-004-37220 Supplier Medchemexpress LLC Supplier No. HY112757100MG
DLinDMA, an ionizable cationic lipid, is a key lipid component of stable nucleic acid lipid particles (SNALPs) as a benchmark. It is used for siRNA delivery. Its structure can be divided into three main regions: the hydrocarbon chains, the linker, and the headgroup.
  • Ionizable cationic lipid
  • Key lipid component of stable nucleic acid lipid particles (SNALPs)
  • Used for siRNA delivery