Learn More
Medchemexpress LLC HY-114331 10mg Medchemexpress, DLK-IN-1 CAS:1620574-24-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11433110MG
Encompass_Preferred
Medchemexpress, HY-114331 10mg DLK-IN-1 CAS:1620574-24-4 DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9329