Learn More
Aviva Systems Biology Corp DLK1 Antibody - middle region (ARP46375_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46375_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DLK1. Alternate names: DLK, FA1, PREF1, Pref-1, ZOG, pG2, Delta1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-59314