Learn More
Enzo Life Sciences DLK1 (human), (recombinant) (His-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX2017640050
Encompass_Preferred
Notch ligand. Alternative name: Fetal antigen 1, Protein delta homolog 1, Preadipocyte factor 1, Pref-1, FA1, pG2. Endotoxin: <0.1ng/µg protein (1 IEU/μg). Purity: ≥95% (SEC-HPLC and reducing SDS-PAGE). Formulation: Lyophilized. In 20mM PBS, pH 7.2, containing 150mM sodium chloride. Source: Produced by HEK 293 cells. DLK1 (Ala24-Pro297) is fused at the C-terminus to a His-tag. Long Term Storage: -20°C. Use/Stability: Lyophilized protein should be stored at < -20°C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7°C for 2-7 days. Aliquots of reconstituted samples are stable at < -20°C for 3 months. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-7986