Learn More
Enzo Life Sciences DLK1 (mouse) monoclonal antibody (PF183E) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX804743C050
Encompass_Preferred
Clone: PF183E. Isotype: IgG2a. Immunogen: Recombinant mouse Dlk1 (extracellular domain). Species reactivity: Mouse. Applications: ELISA, WB. Formulation: Liquid. 0.2μm-filtered solution in PBS, pH 7.4. Contains no preservatives.
Catalog No. 50-200-9968