Learn More
AdipoGen DLK1 mouseFc human rec
SDP
Supplier:  AdipoGen AG40A0107YC010
Encompass_Preferred
Protein. Signal peptide and extracellular domain of mouse DLK1 aa 1-305 are fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02% CHAPS. Purity >95% SDS-PAGE. The transmembrane and secreted protein delta-like 1 homolog DLK1 Protein delta homolog 1 Preadipocyte factor 1 Pref-1 belongs to the EGF-like family. It can be cleaved by a tumor necrosis factor-alpha converting enzyme TACE to release a soluble form. DLK1 plays important roles in regulating cell differentiation such as adipogenesis and osteogenesis. Membrane-bound DLK1 promotes hypertrophic myotube formation, whereas soluble DLK1 inhibits myotube formation. The soluble form of DLK1 inhibits preadipocyte differentiation, while membrane DLK1 represses preadipocyte proliferation. DLK1 is also associated with tumor invasion.
Catalog No. 50-256-2997