Learn More
AdipoGen DLK2 humanFc human rec
SDP
Catalog No. 502562962
Encompass_Preferred
Protein. Signal peptide and the extracellular domain of human DLK2 aa 1-306 are fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. DLK2 regulates adipogenesis.
Catalog No. 50-256-2962 Supplier AdipoGen Supplier No. AG40A0158C050
Protein. Signal peptide and the extracellular domain of human DLK2 aa 1-306 are fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. DLK2 regulates adipogenesis.