Learn More
AdipoGen anti-DLL1 (human), mAb (D1L165-6) (50 µg)
SDP
Supplier:  AdipoGen AG20A0074C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human DLL1. Detects a band of ~68kDa by Western blot. Isotype: Mouse IgG1kappa. Clone: D1L165-6. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. The Notch ligand delta-like protein 1 (DLL1) is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
Catalog No. 50-205-2876