Learn More
AdipoGen DLL1 human pAb
SDP
Supplier:  AdipoGen AG25A0062C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human DLL1. Detects a band of ~68kDa by Western blot. Source Rabbit. Applications ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. The Notch ligand delta-like protein 1 DLL1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
Catalog No. 50-256-3250