Learn More
AdipoGen DLL1 human rec
SDP
Catalog No. 502563015
Encompass_Preferred
Protein. Signal peptide and extracellular domain of human DLL1 aa 1-545 are fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. The Notch ligand delta-like protein 1 DLL1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK-cell precursor.
Catalog No. 50-256-3015 Supplier AdipoGen Supplier No. AG40A0073C050
Protein. Signal peptide and extracellular domain of human DLL1 aa 1-545 are fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. The Notch ligand delta-like protein 1 DLL1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK-cell precursor.