Learn More
AdipoGen DLL1 humanFc human rec
SDP
Catalog No. 502562990
Encompass_Preferred
Protein. Signal peptide and extracellular domain of human DLL1 aa 1-545 are fused at the C-terminus to the Fc portion of human IgG1. Source CHO cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Interacts with human Notch1 as confirmed by flow cytometry. Purity >90% SDS-PAGE. DLL1 Delta-like protein Delta1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK-cell precursor.
Catalog No. 50-256-2990 Supplier AdipoGen Supplier No. AG40A0116YC010
Protein. Signal peptide and extracellular domain of human DLL1 aa 1-545 are fused at the C-terminus to the Fc portion of human IgG1. Source CHO cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Interacts with human Notch1 as confirmed by flow cytometry. Purity >90% SDS-PAGE. DLL1 Delta-like protein Delta1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK-cell precursor.