Learn More
AdipoGen DLL1 mouseFc human rec
SDP
Catalog No. 502562973
Encompass_Preferred
Protein. Extracellular domain of mouse DLL1 aa 18-545 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin <0.1EU/µg. Lyophilized. Contains PBS. The Notch ligand delta-like protein 1 DLL1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK cell precursor.
Catalog No. 50-256-2973 Supplier AdipoGen Supplier No. AG40A0148YC050
Protein. Extracellular domain of mouse DLL1 aa 18-545 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin <0.1EU/µg. Lyophilized. Contains PBS. The Notch ligand delta-like protein 1 DLL1 is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK cell precursor.