Learn More
ABclonal Technology DLL1 Rabbit pAb
SDP
Catalog No. 5000109007
Encompass_Preferred
DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication.
Catalog No. 50-001-09007 Supplier ABclonal Technology Supplier No. A142771000UL
DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication.