Learn More
ACROBiosystems ACROBiosystems Human DLL3 Protein, His Tag
Supplier:  ACROBiosystems DL3H52H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Human DLL3, His Tag (DL3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6872