Learn More
AdipoGen DLL3 (human) (rec.) MultiPack,Protein
SDP
Supplier:  AdipoGen AG40B01513010
Encompass_Preferred
DLL3 (human) (rec.) MultiPack, AdipoGen Life Sciences. Protein. Extracellular domain of human DLL3 (aa 27-466) is fused at the N-terminus to a FLAG?-tag. Source: HEK 293 cells. Endotoxin content: <0.02EU/?g purified protein (LAL test; Lonza). Lyophilized. Contains PBS and CHAPS 0.02%. Purity: >95% (SDS-PAGE). DLL3 (Delta-like protein 3; Delta3) inhibits primary neurogenesis. It plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 binds and activates Notch-1. Defects in DLL3 are the cause of spondylocostal dysostosis type 1 (SCDO1).
Catalog No. 50-101-5657