Learn More
Biomatik Corporation Dll3 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07055
Encompass_Preferred
Dll3 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to DLL3. The Dll3 Recombinant Antibody has been validated for the following applications: ELISA, FC. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dll3 Protein. Applications: ELISA, FC. Target Synonyms: DLL3. Accession Number: Q9NYJ7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9265