Learn More
Aviva Systems Biology Corp DLL4 Antibody - N-terminal region (ARP49436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DLL4. Alternate names: MGC126344, hdelta2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-60084