Learn More
AdipoGen DLL4 (human):Fc (human) (rec.) (highly active mutant) (100 µg)
SDP
Supplier:  AdipoGen AG40B0176C100
Encompass_Preferred
Protein. Extracellular domain of human DLL4 (aa 27-529 plus mutations) are fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). The Notch ligand delta-like protein 4 (DLL4) is expressed highly and selectively within the arterial endothelium and has been shown to function as a ligand for Notch1 and Notch4. It is induced by VEGF as a negative feedback regulator and acts to prevent overexuberant angiogenic sprouting, promoting the timely formation of a well differentiated vascular network. DLL4-Notch1 signaling regulates the formation of appropriate numbers of tip cells to control vessel sprouting and branching in the mouse retina.
Catalog No. 50-205-3276