Learn More
AdipoGen DLL4 humanFc human rec
SDP
Supplier:  AdipoGen AG40A0077YC050
Encompass_Preferred
Protein. Signal peptide and extracellular domain of human DLL4 aa 1-529 are fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.5 % Trehalose. Interacts with human Notch1 as confirmed by flow cytometry. Purity >95% SDS-PAGE. The Notch ligand delta-like protein 4 DLL4 is expressed highly and selectively within the arterial endothelium and has been shown to function as a ligand for Notch1 and Notch4. It is induced by VEGF as a negative feedback regulator and acts to prevent overexuberant angiogenic sprouting, promoting the timely formation of a well differentiated vascular network. DLL4-Notch1 signaling regulates the formation of appropriate numbers of tip cells to control vessel sprouting and branching in the mouse retina.
Catalog No. 50-256-3009