Learn More
AdipoGen DLL4 mouseFc human rec
SDP
Supplier:  AdipoGen AG40A0145C050
Encompass_Preferred
Protein. Signal peptide and extracellular domain of mouse DLL4 aa 1-532 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. The Notch ligand delta-like protein 4 DLL4 is expressed highly and selectively within the arterial endothelium and has been shown to function as a ligand for Notch1 and Notch4. It is induced by VEGF as a negative feedback regulator and acts to prevent overexuberant angiogenic sprouting, pro-moting the timely formation of a well differentiated vascular network. DLL4-Notch1 signaling regulates the formation of appropriate numbers of tip cells to control vessel sprouting and branching in the mouse retina.
Catalog No. 50-256-2975