Learn More
AdipoGen anti-DLL4 (mouse), mAb (9A1.5) (100 µg)
SDP
Supplier:  AdipoGen AG20B0054C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse DLL4. Isotype: Rat IgG1kappa. Clone: 9A1.5. Applications: FACS, ICC. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. The Notch ligand delta-like protein 4 (DLL4) is expressed highly and selectively within the arterial endothelium and has been shown to function as a ligand for Notch1 and Notch4. It is induced by VEGF as a negative feedback regulator and acts to prevent overexuberant angiogenic sprouting, promoting the timely formation of a well differentiated vascular network. DLL4-Notch1 signaling regulates the formation of appropriate numbers of tip cells to control vessel sprouting and branching in the mouse retina.
Catalog No. 50-205-2965