Learn More
Sino Biological Recombinant Human DLL4 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10171H02H500
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Pro524) of human DLL4 (NP_061947.1) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-6426