Learn More
Enzo Life Sciences DLL4 (soluble) (human), (recombinant) (25µg)
SDP
Catalog No. 502007988
Encompass_Preferred
Notch ligand. Alternative name: Delta like protein 4, Delta4. Endotoxin: < 0.1 ng per μg of sDLL-4. Purity: ≥95% (SDS-PAGE and HPLC). MW: ~54kDa. Formulation: Lyophilized. Source: Produced in HEK 293 cells. Recombinant human sDLL-4 contains 498 amino-acid residues. Long Term Storage: -20°C. Use/Stability: The lyophilized protein is stable for at least 2 years from date of receipt at -20°C. Reconstituted sDLL-4 is stable for at least 3 months when stored in working aliquots with a carrier protein at -20°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-7988 Supplier Enzo Life Sciences Supplier No. ALX2017660025
Notch ligand. Alternative name: Delta like protein 4, Delta4. Endotoxin: < 0.1 ng per μg of sDLL-4. Purity: ≥95% (SDS-PAGE and HPLC). MW: ~54kDa. Formulation: Lyophilized. Source: Produced in HEK 293 cells. Recombinant human sDLL-4 contains 498 amino-acid residues. Long Term Storage: -20°C. Use/Stability: The lyophilized protein is stable for at least 2 years from date of receipt at -20°C. Reconstituted sDLL-4 is stable for at least 3 months when stored in working aliquots with a carrier protein at -20°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.