Learn More
ACROBiosystems Recombinant Protein;Human DLL4 Protein, Fc Tag;HEK293;100UG;DL4,DLL4,Delta4,DLL-4,Delta-4
Catalog No. 501658543
Encompass_Preferred
Recombinant Protein;Human DLL4 Protein, Fc Tag;100UG;Human DLL4, Fc Tag (DL4-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # NP_061947.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81 kDa. The protein migrates as 95-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8543 Supplier ACROBiosystems Supplier No. DL4H5259100UG
Recombinant Protein;Human DLL4 Protein, Fc Tag;100UG;Human DLL4, Fc Tag (DL4-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # NP_061947.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81 kDa. The protein migrates as 95-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.