Learn More
ACROBiosystems Recombinant Protein;Human DLL4 Protein;HEK293;1MG;DL4,DLL4,Delta4,DLL-4,Delta-4
Catalog No. 501658591
Encompass_Preferred
Recombinant Protein;Human DLL4 Protein;1MG;Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # AAI06951).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.1 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8591 Supplier ACROBiosystems Supplier No. DL4H52271MG
Recombinant Protein;Human DLL4 Protein;1MG;Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # AAI06951).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.1 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.