Learn More
Echelon Biosciences Research Labs DLPE 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L2112250MG
Encompass
1,2-Dilauroyl-sn-glycero-3-phosphoethanolamine (DLPE) is a water soluble derivative of phosphatidylethanolamine with lauric acid (12:0) acyl chains. Phosphatidylethanolamine (PE) is formed primarily in the reaction of CDP-ethanolamine and diacylglycerol. PE is one of the major components of cell membranes in bacteria and in nervous system. This item is non returnable
Catalog No. 50-720-499