Learn More
Cayman Chemical DL-erythro-PPMP (hydrochloride), Purity: ≥98%, 100 mg
SDP
Catalog No. 503015985
Encompass_Preferred
A stereoisomer of DL-threo-PPMP; has no effect on glucosylceramide synthetase activity; used as a negative control for DL-threo-PPMP, D-threo-PPMP, and L-threo-PPMP inhibitory activity
Catalog No. 50-301-5985 Supplier Cayman Chemical Supplier No. 24524100 MG
A stereoisomer of DL-threo-PPMP; has no effect on glucosylceramide synthetase activity; used as a negative control for DL-threo-PPMP, D-threo-PPMP, and L-threo-PPMP inhibitory activity