Learn More
Medchemexpress LLC DLST protein, human (Sf9, His) | >90.0% | 10 UG
SDP
Catalog No. 5000294344
Encompass_Preferred
DLST is a recombinant human dihydrolipoamide succinyltransferase fragment expressed in Sf9 insect cells with an N-terminal His tag. It functions as part of the 2-oxoglutarate dehydrogenase complex, catalyzing conversion of 2-oxoglutarate to succinyl-CoA and CO2; the protein is mitochondrial and contributes to histone succinylation by supplying succinyl-CoA to succinyltransferases.
  • Expressed in Sf9 insect cells for eukaryotic folding and modifications.
  • N-terminal His tag to facilitate affinity purification.
  • Residues D68-L453 representing the mature protein fragment.
  • Approximate molecular weight 43.9 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Formulated in 20 mM Tris, 300 mM NaCl, pH 7.5, 10% glycerol, 0.22 μm filtered.
  • Endotoxin level <1 EU/μg and shipped on dry ice.
  • Available in small research quantities (for example, 10 μg).
Catalog No. 50-002-94344 Supplier Medchemexpress LLC Supplier No. HYP7570910UG
DLST is a recombinant human dihydrolipoamide succinyltransferase fragment expressed in Sf9 insect cells with an N-terminal His tag. It functions as part of the 2-oxoglutarate dehydrogenase complex, catalyzing conversion of 2-oxoglutarate to succinyl-CoA and CO2; the protein is mitochondrial and contributes to histone succinylation by supplying succinyl-CoA to succinyltransferases.
  • Expressed in Sf9 insect cells for eukaryotic folding and modifications.
  • N-terminal His tag to facilitate affinity purification.
  • Residues D68-L453 representing the mature protein fragment.
  • Approximate molecular weight 43.9 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Formulated in 20 mM Tris, 300 mM NaCl, pH 7.5, 10% glycerol, 0.22 μm filtered.
  • Endotoxin level <1 EU/μg and shipped on dry ice.
  • Available in small research quantities (for example, 10 μg).