Learn More
Cayman Chemical DLSulforaphn NacetylLcy 10mg
SDP
Catalog No. 502971164
Encompass_Preferred
A major metabolite of sulforaphane that has been shown induce ARE-driven gene expression with roughly 8-fold less potency than sulforaphane
Catalog No. 50-297-1164 Supplier Cayman Chemical Supplier No. 1609810 MG
A major metabolite of sulforaphane that has been shown induce ARE-driven gene expression with roughly 8-fold less potency than sulforaphane