Learn More
Cayman Chemical DL-threo-PDMP (hydrochloride), Purity: ≥98%, CAS: 80938-69-8, 100 mg
SDP
Catalog No. 502890754
Encompass_Preferred
An inhibitor of sphingolipid biosynthesis; inhibits glucosylceramide synthase by 33 and 48% in MDCK cell homogenates at 5 and 10 µM; reduces the synthesis of glucosylceramide increases cellular ceramide, and induces cell cycle arrest; increases Aβ42 and Aβ39 production independent of ceramide metabolism via modulation of γ-secretase activity
Catalog No. 50-289-0754 Supplier Cayman Chemical Supplier No. 10005276100 MG
An inhibitor of sphingolipid biosynthesis; inhibits glucosylceramide synthase by 33 and 48% in MDCK cell homogenates at 5 and 10 µM; reduces the synthesis of glucosylceramide increases cellular ceramide, and induces cell cycle arrest; increases Aβ42 and Aβ39 production independent of ceramide metabolism via modulation of γ-secretase activity