Learn More
Cayman Chemical DL-threo-PPMP (hydrochloride), Purity: ≥98%, CAS: 139974-41-7, 100 mg
SDP
Catalog No. 502966512
Encompass_Preferred
An inhibitor of glucosylceramide synthase; inhibits glucosylceramide synthase by 70, 41, and 62% in MDCK cell homogenates, mouse liver microsomes, and mouse brain homogenates, respectively, at 20 µM; inhibits sphingomyelin synthase activity in P. falciparum-infected erythrocytes and inhibits growth of P. falciparum (IC50 = 0.85 µM)
Catalog No. 50-296-6512 Supplier Cayman Chemical Supplier No. 17236100 MG
An inhibitor of glucosylceramide synthase; inhibits glucosylceramide synthase by 70, 41, and 62% in MDCK cell homogenates, mouse liver microsomes, and mouse brain homogenates, respectively, at 20 µM; inhibits sphingomyelin synthase activity in P. falciparum-infected erythrocytes and inhibits growth of P. falciparum (IC50 = 0.85 µM)