Learn More
Cayman Chemical DL-Tryptophan octyl ester (hydrochloride), Purity: ≥98%, CAS: 6278-90-6, 500 mg
SDP
Catalog No. 502965626
Encompass_Preferred
A stable, cell-permeable tryptophan derivative with an octyl ester protected COOH group
Catalog No. 50-296-5626 Supplier Cayman Chemical Supplier No. 17599500 MG
A stable, cell-permeable tryptophan derivative with an octyl ester protected COOH group